Imprimer
Graphite electrodes, counter
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
14746.KN 14751.KN 14754.KN
14746.KNEA 393 EUR
14746.KN 14751.KN 14754.KN
Graphite electrodes, counter
Électrodes Électrodes, autres
Graphite electrodes are used as homogeneous catalysts.
  • AGKSP grade
  • Ultra F purity
  • Light sensitive and hygroscopic
  • Incompatible with oxidising agents
  • Applications include piston rings, thrust bearings, journal bearings and vanes
These are widely used in electric arc furnaces (EAF) and ladle furnaces (LF) for the melting of steel, non-ferrous metals and some non-metallic minerals. Carbon based seals are used in the shafts and fuel pumps of many aircraft jet engines.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable