Imprimer

Sodium nitrate ≥99.0% ACS

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

14493.14 14493.30 14493.A1 14493.A7
14493.14EA 41.4 EUR
14493.14 14493.30 14493.A1 14493.A7
Sodium nitrate ≥99.0% ACS
Sodium nitrate

In manufacturing of HNO₃, as a catalyst to manufacture H₂SO₄, in manufacturing of glass, enamel for pottery, sodium nitrite


Purity: 99.0% min CAS No.: 7631-99-4 Molecular Formula: NaNO3 Formula Weight: 84.99 Physical Form: Crystalline Density: 2.26 Appearance: White Odour: Odourless Melting Point: 308° Solubility: Soluble in water. Slightly soluble in methanol Merck Reference: 14,8647 UN No.: 1498 MDL No.: MFCD00011119


In manufacturing of HNO3, as a catalyst to manufacture H2SO4, in manufacturing of glass, enamel for pottery, sodium nitrite

Formule: NaNO₃
Poids moléculaire: 84.99 g/mol
Point d'ébullition: 380
Point de fusion: 308
Densité: 2.26
Température de stockage: Température ambiante
Numéro MDL: MFCD00011119
Numéro CAS: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1 III
REACH: 01-2119488221-41
Indice Merck: 14,8647

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR