Imprimer

Calcium standard solution, 10,000 mg/L Ca in 5% nitric acid (from CaCO₃) Specpure® étalon mono-élément pour ICP

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

14407.AD 14407.AE 14407.AP
14407.ADEA 126 EUR
14407.AD 14407.AE 14407.AP
Calcium standard solution, 10,000 mg/L Ca in 5% nitric acid (from CaCO₃) Specpure® étalon mono-élément pour ICP
Étalons Étalons cationiques Standards de calcium

Molecular Formula: CaCO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085314


Certifications: Traçabilité NIST

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR