Imprimer

Nickel standard dissolved in oil, 1,000 µg/g Ni in mineral oil Specpure® for wear metal analysis

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

14211.18
14211.18EA 112 EUR
14211.18
Nickel standard dissolved in oil, 1,000 µg/g Ni in mineral oil Specpure® for wear metal analysis
Étalons Étalons cationiques Standards de nickel

Physical Form: Liquid
MDL No.: MFCD00011137

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR