Imprimer
Lipid Removal Agent (LRA), Pore size: 90 Å, powder 325 mesh, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
13358-U 13360-U
13358-UEA 142 EUR
13358-U 13360-U
Lipid Removal Agent (LRA), Pore size: 90 Å, powder 325 mesh, Supelco®
Réactifs pour acides nucléiques Nucléotides
Lipid removal agent is a synthetic adsorbent of crystalline calcium silicate hydrate which is used for removing lipids and endotoxin from plasma or aqueous solutions. It is also used for DNA purification to remove protein, RNA and genomic DNA.
  • Surface area: 120 m²/g
  • matrix active group: Silica
  • NACRES SB.52
Lipid removal agent (LRA) was used for absorption of lipids using gel permeation chromatography.
LRA was used to isolate lipoprotein particles from coeluting proteins in collected fraction from Gel filtration chromatography.