Imprimer
Telescopic handles
Fournisseur: FHP VILEDA SCS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
119966 111380
129-0266EA 38.9 EUR
129-0266 129-0456
Telescopic handles
Serpillières
These lightweight, strong extendable handles are perfect for all controlled environments. Fully autoclavable.
  • Extremely easy to use and extend with a simple twist of the wrist
  • Resist autoclave, gamma and ETO processes
  • Ergonomic center and anti-slip grips
  • Available with colour coding clips (Blue, Red, Yellow, Green)
For use with all Vileda Professional frames and CE Squeegee.