Imprimer
European earth bonding plugs
Fournisseur: DESCO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
231135
115-3043EA 0 EUR
115-3043
European earth bonding plugs
Prises électriques
These plugs to fit UK European CEE (7) VII Schuko sockets as used in Belgium, France, Germany, Holland, Spain. The Europe Earth Bonding Point (EBP) plug is designed to provide a common ground point for grounding using protective earth in an ESD protected area (EPA). The plug fits into the mains supply socket, making a connection with the earth conductor only. In place of the live and neutral pins are moulded insulating plastic pins to allow positive location in the socket. Connectors on the front of the plug are available for connection via ground cords to the various elements of the EPA.En savoir plus