Imprimer
Cleanroom wipes, ReFIBE™
Fournisseur: CONTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
RFHS-1212 RFHS-99 RFHS-1212IR RFHS-99IR
115-2510EA 1080 EUR
115-2510 115-2511 115-3074 115-2679
Cleanroom wipes, ReFIBE™
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
Recycled 100% knitted polyester cleanroom wipes offer a sustainable solution for cleanroom environments, without jeopardising quality or performance. Soft textured wipes clean surfaces without scratching, while offering low levels of particles and fibres, abrasion and chemical resistance, and good solvent compatibility.
  • 100% knitted laundered polyester
  • Laser cut sealed edges seals fibres at wipe's edge to reduce shedding
  • Suitable for use in high grade cleanrooms
  • Manufactured from 100% recycled plastic
Each wipe is constructed from 100% post-consumerEn savoir plus