Imprimer
Lingettes pour salles blanches, Polynit Heatseal
Fournisseur: CONTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
PNHS-1212 PNHS-44 PNHS-99 PNHS-99B/150 PNHS-66 LWPS0030 PNHS-99B PNHS-44B LWPN0020
115-0375EA 111 EUR
115-0375 115-0376 115-0268 CONTPNHS-99B/150 115-2148 CONTLWPS0030 CONTPNHS-99B CONTPNHS-44B CONTLWPN0020
Lingettes pour salles blanches, Polynit Heatseal
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
Ideal for wiping exterior surfaces of products and cleaning interior surfaces of process tools or other equipment. Made from 100% textured polyester no-run knit.
  • Good solvent sorbency, abrasion and chemical resistance
  • Extremely low in particles and fibres
  • Laser cutting seals fibres for non abrasive edges
Suitable for use in Class 1, ISO 3, M1.5 cleanrooms
Colour: White.