Imprimer
ESD safety shoes, lace-up, uvex 1 sport S1 SRC
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
6598.8/38 6598.8/39 6598.8/40 6598.8/41 6598.8/42 6598.8/43 6598.8/44 6598.8/45 6598.8/46 6598.8/47 6598.8/48 6598.8/49 6598.8/50 6598.8/51 6598.8/52 6598.8/35 6598.8/36 6598.8/37 6598.7/37 6598.7/38 6598.7/49 6598.7/47 6598.7/52 6598.7/45 6598.7/39 6598.7/41 6598.7/44 6598.7/35 6598.7/36 6598.7/40 6598.7/42 6598.7/43 6598.7/48 6598.7/51 6598.7/46 6598.7/50 6598.9/35 6598.9/37 6598.9/36 6598.9/40 6598.9/42 6598.9/44 6598.9/47 6598.9/51 6598.9/43 6598.9/45 6598.9/49 6598.9/50 6598.9/38 6598.9/46 6598.9/52 6598.9/39 6598.9/41 6598.9/48
114-8602EA 114 EUR
114-8602 114-8603 114-8604 114-8605 114-8606 114-8607 114-8608 114-8609 114-8610 114-8611 114-8612 114-8613 114-8614 114-8615 114-8616 114-8599 114-8600 114-8601 114-8583 114-8584 114-8595 114-8593 114-8598 114-8591 114-8585 114-8587 114-8590 114-8581 114-8582 114-8586 114-8588 114-8589 114-8594 114-8597 114-8592 114-8596 114-8617 114-8619 114-8618 114-8622 114-8624 114-8626 114-8629 114-8633 114-8625 114-8627 114-8631 114-8632 114-8620 114-8628 114-8634 114-8621 114-8623 114-8630
ESD safety shoes, lace-up, uvex 1 sport S1 SRC
Chaussures et bottes
Metal-free, extremely lightweight, flexible, multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, plus a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Breathable distance mesh lining
  • Anti-slip sole with shock-absorbent heel and front of the foot
  • Virtually seamless design eliminates potential pressure points and
En savoir plus