Imprimer
Gants résistants aux produits chimiques, Powercoat® 950-20 Neofit
Fournisseur: HONEYWELL SÉCURITÉ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
2095020/7 2095020/8 2095020/9 2095020/10 2095020/11
112-4040EA 5.3 EUR
112-4040 112-4041 112-4042 112-4043 112-4044
Gants résistants aux produits chimiques, Powercoat® 950-20 Neofit
Gants
This durable glove made from neoprene provides protection against a wide range of chemical products (detergents, some solvents, acids and bases).
  • Neoprene provides chemical and mechanical resistance
  • Diamond finish provides a better grip in damp environments
  • Cotton flock interior makes it easier to put on and off the glove
  • Long cuff helps to protect a large part of the forearm
Ideal for battery handling but can also be used to handle certain dilute acids.
Thickness: 0,72 mm
Colour: Black
EN 374 (AKL), EN 388 (3111), Cat. III
Certifications:En savoir plus