Imprimer

Amberlite® XAD4, Pore dim.: 100 Å 20 - 60 mesh resin_ion-exchange, Supelco®

Marques : Merck
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

10358
10358.EA 183 EUR
10358.
Amberlite® XAD4, Pore dim.: 100 Å 20 - 60 mesh resin_ion-exchange, Supelco®
Amberlite® XAD4

Amberlite XAD4, a nonpolar absorbent, used for stripping and concentration of organic compounds. It is also used in developing chelating ligands due to its properties like, porosity, high surface area, purity and durability.


  • Surface area: 750 m²/g
  • Used in rapid fractionation technique
  • Used as sorbent for HPLC analysis


Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.


Polyaromatic adsorbent for small hydrophobic compounds, surfactants, pharmaceutical manufacturing, phenols, chlorinated organics, pesticide removal and recovery, and organic removal from aqueous food streams.

Numéro MDL: MFCD00132704
Numéro CAS: 9003-69-4

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR