Imprimer

Hydroxylammonium chlorure ACS, ISO analytical reagent, Supelco®

Marques : Merck
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Attention

Synonyme(s): Hydroxylamine chlorhydrate

MQ100

1.04619.0250
1.04619.0250EA 432 EUR
1.04619.0250
Hydroxylammonium chlorure ACS, ISO analytical reagent, Supelco®
Hydroxylammonium chlorure
Formule: NH₂OH·HCl
Poids moléculaire: 69.49 g/mol
Point d'ébullition: 306
Point de fusion: 151
Densité: 1.67 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00051089
Numéro CAS: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8 III
Indice Merck: 13,04853

Commander maintenant

Résultats des tests de spécification

Assay (manganometric) ≥ 99.0 %
Clarity of the solution in Ethanol passes test
pH-value (5 %; water) 2.5 - 3.5
Titrable free acid ≤ 0.25 meq/g
Sulfate (SO₄) ≤ 0.002 %
Heavy metals (ACS) ≤ 0.0005 %
Cu (Copper) ≤ 0.001 %
Fe (Iron) ≤ 0.0005 %
Hg (Mercury) ≤ 0.000001 %
NH₄ (Ammonium) passes test
Pb (Lead) ≤ 0.0005 %
Total sulfur (as sulfate) ≤ 0.005 %
Residue on ignition (as sulfate) ≤ 0.01 %
Corresponds to ACS,ISO


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR