Imprimer

n-Hexane, Uvasol® pour la spectroscopie, Supelco®

Marques : Merck
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): Hexyl hydride, Hexanen, Hexane

M-Clarity™ (Niveau de Qualité) = MQ100

1.04372.0500 1.04372.2500
1.04372.0500EA 157 EUR
1.04372.0500 1.04372.2500
n-Hexane, Uvasol® pour la spectroscopie, Supelco®
n-Hexane
Formule: H₃C(CH₂)₄CH₃
Poids moléculaire: 86.18 g/mol
Point d'ébullition: 69
Point de fusion: –94.3 °C
Densité: 0.659 g/cm³ (20 °C)
Point éclair: –22 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00009520
Numéro CAS: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Indice Merck: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Purity (GC) ≥99.0%
Evaporation Residue ≤0.0002%
Water ≤0.005%
Color ≤10 Hazen
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0002 meq/g
Fluorescence (as quinine at 254 nm) ≤1.0 ppb
Fluorescence (as quinine at 365 nm) ≤1.0 ppb
Transmission (at 195 nm) ≥10%
Transmission (at 210 nm) ≥60%
Transmission (at 217 nm) ≥80%
Transmission (at 225 nm) ≥90%
Transmission (from 245 nm) ≥98%
Absorbance (at 195 nm) ≤1.00
Absorbance (at 210 nm) ≤0.22
Absorbance (at 217 nm) ≤0.10
Absorbance (at 225 nm) ≤0.05
Absorbance (from 245 nm) ≤0.01
Meet the requirements of 'Specific use spectroscopy' according Reag. Ph. Eur.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR