Imprimer

Propanol-2, LiChrosolv®, gradient LC, Supelco®

Marques : Merck
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): Diméthylcarbinol, IPA, Alcool isopropylique, Isopropanol

MQ100

1.01040.1000 1.01040.2500 1.01040.4000 1.01040.9030 1.01040.9185
1.01040.1000EA 50.35 EUR
1.01040.1000 1.01040.2500 1.01040.4000 1.01040.9030 1.01040.9185
Propanol-2, LiChrosolv®, gradient LC, Supelco®
Propanol-2

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Formule: (CH₃)₂CHOH
Poids moléculaire: 60.1 g/mol
Point d'ébullition: 82
Point de fusion: –89 °C
Densité: 0.786 g/cm³ (20 °C)
Point éclair: 12
Température de stockage: Température ambiante
Numéro MDL: MFCD00011674
Numéro CAS: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Indice Merck: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR