Imprimer

Acide chlorhydrique 25%, EMSURE® analysis_for, Supelco®

Marques : Merck
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): Hydrochloric acid solution, Muriatic acid

MQ300

1.00316.1000 1.00316.1011 1.00316.2500 1.00316.2511 1.00316.9025 1.00316.9950
1.00316.1000EA 26.47 EUR
1.00316.1000 1.00316.1011 1.00316.2500 1.00316.2511 1.00316.9025 1.00316.9950
Acide chlorhydrique 25%, EMSURE® analysis_for, Supelco®
Acide chlorhydrique

For full details of specification and hazards please see www.vwr.com or the VWR Chemicals catalogues

Formule: HCl
Poids moléculaire: 36.46 g/mol
Point d'ébullition: 107 °C (1013 hPa)
Point de fusion: –52 °C
Densité: 1.12 g/cm3 (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00011324
Numéro CAS: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Commander maintenant

Résultats des tests de spécification

Assay (Alkalimetric) ≥25.0%
Color ≤10 Hazen
Bromide (Br) ≤50 ppm
Free Chlorine (Cl) ≤0.5 ppm
Sulfate (SO₄) ≤1 ppm
Sulfite (SO₃) ≤1 ppm
Al (Aluminium) ≤0.050 ppm
As (Arsenic) ≤0.01 ppm
Ba (Barium) ≤0.020 ppm
Be (Beryllium) ≤0.020 ppm
Bi (Bismuth) ≤0.100 ppm
Ca (Calcium) ≤0.500 ppm
Cd (Cadmium) ≤0.010 ppm
Co (Cobalt) ≤0.010 ppm
Cr (Chromium) ≤0.020 ppm
Cu (Copper) ≤0.020 ppm
Fe (Iron) ≤0.200 ppm
Ge (Germanium) ≤0.050 ppm
K (Potassium) ≤0.100 ppm
Li (Lithium) ≤0.010 ppm
Mg (Magnesium) ≤0.100 ppm
Mn (Manganese) ≤0.010 ppm
Mo (Molybdenum) ≤0.020 ppm
NH₄ (Ammonium) ≤1 ppm
Na (Sodium) ≤0.500 ppm
Ni (Nickel) ≤0.020 ppm
Pb (Lead) ≤0.020 ppm
Sr (Strontium) ≤0.010 ppm
Ti (Titanium) ≤0.100 ppm
Tl (Thallium) ≤0.050 ppm
V (Vanadium) ≤0.010 ppm
Zn (Zinc) ≤0.050 ppm
Zr (Zirconium) ≤0.100 ppm
Residue on Ignition (as SO₄) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR