Imprimer

Bismuth (III) carbonate basique Blend ZS, Macron Fine Chemicals™

Marques : Avantor
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): Dioxycarbonate de dibismuth

0288-24EA 0 EUR
0288-24
Bismuth (III) carbonate basique Blend ZS, Macron Fine Chemicals™
Bismuth (III) carbonate basique

Bismuth Subcarbonate

Formule: (BiO)₂CO₃
Poids moléculaire: 509.97 g/mol
Densité: 6.86 kg/L
Température de stockage: Température ambiante
Numéro MDL: MFCD00010877
Numéro CAS: 5892-10-4
EINECS: 215-185-5
REACH: 01-2119457892-27
Indice Merck: 13,01283

Commander maintenant

Résultats des tests de spécification

For Laboratory,Research,or Manufacturing Use Passes Test
Appearance (white to off-white powder) Passes Test
pH 5.0 - 8.5
Identification (Zinc Stearate) Passes Test
Loss on Ignition, For Information Only For Information Only
Bismuth Assay, For Information Only
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR