Imprimer
Amberlite® IR120 Hydrogen form, env. 50% d'humidité 620 - 830 μm (échangeur de cations fortement acide), résine échangeuse d'anions, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
06428-1KG 06428-5KG
06428-1KGEA 101 EUR
06428-1KG 06428-5KG
Amberlite® IR120 Hydrogen form, env. 50% d'humidité 620 - 830 μm (échangeur de cations fortement acide), résine échangeuse d'anions, Supelco®
Amberlite® IR120 Hydrogen form
Amberlite IR120 is a gel-type strongly acidic cation exchange resin and a heterogeneous catalyst, suitable for a wide variety of chemical process applications, removal of amino acids at low pH, USP potassium methods, etc..
Amberlite IR120 has been used as a catalyst support to form chelates with Co2+ or Ru3+ ions for subsequent hydrogen generation by hydrolysis of ammonia borane. It may be used as a catalyst for the oxirane ring-opening reaction effected by hydrogen peroxide (H2O2) during the synthesis of epoxides from triglycerides source. Amberlite IR120 hydrogen form may be used to investigate the ion-exchange equilibria of H+/Pb2+, H+/Ni2+, and H+/Cr3+ systems.
Numéro MDL: MFCD00132707
Numéro CAS: 39389-20-3