Bromure d'ammonium ≥99.0% ACS
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
Synonyme(s):
Ammonium bromure
Purity: 99.0% min
CAS No.: 12124-97-9
Molecular Formula: NH4Br
Formula Weight: 97.97
Physical Form: Granular
Density: 2.429
Appearance: White
Melting Point: 452° subl.
Solubility: Freely soluble in water, methanol, ethanol, and acetone. Slightly soluble in ether. Practically insoluble in ethyl acetate
Merck Reference: 14,505
MDL No.: MFCD00011419
Corrosion inhibitors, manufacturing of photographic films, plates, paper, lithography, fireproofing of woods
Formule:
H₄BrN Point de fusion: 452 °C (subl.) Densité: 2,429 Température de stockage: Température ambiante |
Numéro MDL:
MFCD00011419 Numéro CAS: 12124-97-9 EINECS: 235-183-8 Indice Merck: 14,505 |